trans-3-Fluoro-tetrahydro-2H-pyran-4-yl methanesulphonate racemate

Lieferant: Apollo Scientific

PC430262-1G
APOSPC430262-1GEA 865 EUR
APOSPC430262-1G
trans-3-Fluoro-tetrahydro-2H-pyran-4-yl methanesulphonate racemate
trans-3-Fluoro-tetrahydro-2H-pyran-4-yl methanesulphonate racemate
Formel: C₆H₁₁FO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD28348444
CAS-Nummer: 1864003-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR