3-Cyclopropyl-4-fluoroaniline

Lieferant: Apollo Scientific
Achtung

PC430268-1G PC430268-250MG PC430268-5G
APOSPC430268-1GEA 0 EUR
APOSPC430268-1G APOSPC430268-250MG APOSPC430268-5G
3-Cyclopropyl-4-fluoroaniline
3-Cyclopropyl-4-fluoroaniline
Formel: C₉H₁₀FN
Lagertemperatur: Gefrierschrank
MDL: MFCD19443457
CAS-Nummer: 890129-90-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR