tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate

PC430278-1G
APOSPC430278-1GEA 865 EUR
APOSPC430278-1G
tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate
tert-Butyl-4-(5-chloro-2-fluorobenzoyl)piperidine-1-carboxylate
Formel: C₁₇H₂₁ClFNO₃
Molecular Weight: 341,81 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16618544
CAS-Nummer: 1228631-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR