tert-Butyl 4-(5-bromo-2-fluorobenzoyl)piperidine-1-carboxylate
Lieferant: Apollo Scientific
PC430279-1G
APOSPC430279-1GEA
865
EUR
APOSPC430279-1G
tert-Butyl 4-(5-bromo-2-fluorobenzoyl)piperidine-1-carboxylate
tert-Butyl 4-(5-bromo-2-fluorobenzoyl)piperidine-1-carboxylate
Formel:
C₁₇H₂₁BrFNO₃ Molecular Weight: 386,256 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16618546 CAS-Nummer: 1228631-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...