tert-Butyl 4-(2,5-difluorobenzoyl)piperidine-1-carboxylate
Lieferant: Apollo Scientific
PC430280-1G
APOSPC430280-1GEA
568
EUR
APOSPC430280-1G
tert-Butyl 4-(2,5-difluorobenzoyl)piperidine-1-carboxylate
tert-Butyl 4-(2,5-difluorobenzoyl)piperidine-1-carboxylate
Formel:
C₁₇H₂₁F₂NO₃ Molecular Weight: 325,35 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16618547 CAS-Nummer: 1228631-03-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...