1-Benzyl-3-Fluoropiperidin-4-one hydrochloride

Lieferant: Apollo Scientific

PC430282-1G
APOSPC430282-1GEA 655 EUR
APOSPC430282-1G
1-Benzyl-3-Fluoropiperidin-4-one hydrochloride
1-Benzyl-3-Fluoropiperidin-4-one hydrochloride
Formel: C₁₂H₁₄FNO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27978592
CAS-Nummer: 1864064-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR