1-Benzyl-3,3-difluoropiperidin-4-ol

Lieferant: Apollo Scientific

PC430295-1G
APOSPC430295-1GEA 764 EUR
APOSPC430295-1G
1-Benzyl-3,3-difluoropiperidin-4-ol
1-Benzyl-3,3-difluoropiperidin-4-ol
Formel: C₁₂H₁₅F₂NO
Molecular Weight: 227,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18433631
CAS-Nummer: 1239596-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR