Benzyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate

Lieferant: Apollo Scientific

PC430296-1G
APOSPC430296-1GEA 865 EUR
APOSPC430296-1G
Benzyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate
Benzyl 3,3-difluoro-4-hydroxypiperidine-1-carboxylate
Formel: C₁₃H₁₅F₂NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD16250752
CAS-Nummer: 1255666-33-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR