tert-Butyl 4-amino-3,3-difluoropiperidine-1-carboxylate 95+%

Lieferant: Apollo Scientific

PC430297-250MG
APOSPC430297-250MGEA 58 EUR
APOSPC430297-250MG
tert-Butyl 4-amino-3,3-difluoropiperidine-1-carboxylate 95+%
tert-Butyl 4-amino-3,3-difluoropiperidine-1-carboxylate
Formel: C₁₀H₁₈F₂N₂O₂
Molecular Weight: 236,259 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16250242
CAS-Nummer: 1255666-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR