2-(Difluormethoxy)benzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC4303-5G
PC4303-25G
PC4303-100G
APOSPC4303-5GEA
37.7
EUR
APOSPC4303-5G
APOSPC4303-25G
APOSPC4303-100G
2-(Difluormethoxy)benzylalkohol 98%
2-(Difluormethoxy)benzylalkohol
Formel:
C₈H₈F₂O₂ Schmelzpunkt: 42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00236226 CAS-Nummer: 72768-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...