tert-Butyl 4-hydroxy-4-(trifluoromethyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

PC430306-1G
APOSPC430306-1GEA 655 EUR
APOSPC430306-1G
tert-Butyl 4-hydroxy-4-(trifluoromethyl)piperidine-1-carboxylate
tert-Butyl 4-hydroxy-4-(trifluoromethyl)piperidine-1-carboxylate
Formel: C₁₁H₁₈F₃NO₃
Molecular Weight: 269,261 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16990768
CAS-Nummer: 550371-74-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR