Benzyl 3,3-difluoro-4-oxopiperidine-1-carboxylate

Lieferant: Apollo Scientific

PC430307-1G
APOSPC430307-1GEA 865 EUR
APOSPC430307-1G
Benzyl 3,3-difluoro-4-oxopiperidine-1-carboxylate
Benzyl 3,3-difluoro-4-oxopiperidine-1-carboxylate
Formel: C₁₃H₁₃F₂NO₃
Molecular Weight: 269,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16250753
CAS-Nummer: 1283720-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR