1-(Trifluoroacetyl)piperidine-4-carboxylic acid
Lieferant: Apollo Scientific
Synonyme:
1-(2,2,2-Trifluoroacetyl)-4-piperidinecarboxylic acid, 1-(2,2,2-Trifluoroacetyl)piperidine-4-carboxylic acid
PC430312-1G
APOSPC430312-1GEA
109
EUR
APOSPC430312-1G
1-(Trifluoroacetyl)piperidine-4-carboxylic acid
1-(Trifluoroacetyl)piperidine-4-carboxylic acid
Formel:
C₈H₁₀F₃NO₃ Molecular Weight: 225,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03840837 CAS-Nummer: 126501-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...