2,2,2-Trifluoro-1-(4-(4-fluorobenzoyl)piperidin-1-yl)ethanone
Lieferant: Apollo Scientific
PC430316-1G
APOSPC430316-1GEA
263
EUR
APOSPC430316-1G
2,2,2-Trifluoro-1-(4-(4-fluorobenzoyl)piperidin-1-yl)ethanone
2,2,2-Trifluoro-1-(4-(4-fluorobenzoyl)piperidin-1-yl)ethanone
Formel:
C₁₄H₁₃F₄NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11036250 CAS-Nummer: 1159982-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...