tert-Butyl-4-((4-fluorophenyl)(hydroxy)methyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

PC430317-1G
APOSPC430317-1GEA 364 EUR
APOSPC430317-1G
tert-Butyl-4-((4-fluorophenyl)(hydroxy)methyl)piperidine-1-carboxylate
tert-Butyl-4-((4-fluorophenyl)(hydroxy)methyl)piperidine-1-carboxylate
Formel: C₁₇H₂₄FNO₃
Molecular Weight: 309,38 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11041269
CAS-Nummer: 160296-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR