4-Fluoromethylpiperidine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Fluoromethyl)piperidine hydrochloride

PC430318-1G
APOSPC430318-1GEA 764 EUR
APOSPC430318-1G
4-Fluoromethylpiperidine hydrochloride
4-Fluoromethylpiperidine hydrochloride
Formel: C₆H₁₃ClFN
Molecular Weight: 153,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19443952
CAS-Nummer: 787564-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR