1-N-Boc-4-Fluoromethylpiperidine
Lieferant: Apollo Scientific
PC430319-1G
APOSPC430319-1GEA
764
EUR
APOSPC430319-1G
1-N-Boc-4-Fluoromethylpiperidine
1-N-Boc-4-Fluoromethylpiperidine
Formel:
C₁₁H₂₀FNO₂ Molecular Weight: 217,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11054100 CAS-Nummer: 259143-03-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...