tert-Butyl-3-(trifluoromethyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

PC430321-1G
APOSPC430321-1GEA 865 EUR
APOSPC430321-1G
tert-Butyl-3-(trifluoromethyl)piperidine-1-carboxylate
tert-Butyl-3-(trifluoromethyl)piperidine-1-carboxylate
Formel: C₁₁H₁₈F₃NO₂
Molecular Weight: 253,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054108
CAS-Nummer: 1093759-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR