1-N-Boc-4-Fluoropiperidine

Lieferant: Apollo Scientific

PC430322-1G
APOSPC430322-1GEA 865 EUR
APOSPC430322-1G
1-N-Boc-4-Fluoropiperidine
1-N-Boc-4-Fluoropiperidine
Formel: C₁₀H₁₈FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11054109
CAS-Nummer: 178181-55-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR