1-Benzyl-3-(2,2,2-trifluoroacetyl)piperidin-4-one hydrochloride

Lieferant: Apollo Scientific

PC430324-1G
APOSPC430324-1GEA 568 EUR
APOSPC430324-1G
1-Benzyl-3-(2,2,2-trifluoroacetyl)piperidin-4-one hydrochloride
1-Benzyl-3-(2,2,2-trifluoroacetyl)piperidin-4-one hydrochloride
Formel: C₁₄H₁₄F₃NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD11840353
CAS-Nummer: 1198285-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR