1,1'-(Piperidine-1,3-diyl)bis(2,2,2-trifluoroethanone)

Lieferant: Apollo Scientific

PC430326-1G
APOSPC430326-1GEA 655 EUR
APOSPC430326-1G
1,1'-(Piperidine-1,3-diyl)bis(2,2,2-trifluoroethanone)
1,1'-(Piperidine-1,3-diyl)bis(2,2,2-trifluoroethanone)
Formel: C₉H₉F₆NO₂
Molecular Weight: 277,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11840380
CAS-Nummer: 1159982-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR