1-(1-Benzylpiperidin-4-yl)-2,2,2-trifluoroethanone oxalate

Lieferant: Apollo Scientific

PC430327-1G
APOSPC430327-1GEA 655 EUR
APOSPC430327-1G
1-(1-Benzylpiperidin-4-yl)-2,2,2-trifluoroethanone oxalate
1-(1-Benzylpiperidin-4-yl)-2,2,2-trifluoroethanone oxalate
Formel: C₁₆H₁₈F₃NO₅
Lagertemperatur: Raumtemperatur
MDL: MFCD12026432
CAS-Nummer: 1198286-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR