tert-Butyl 4-(2,2,2-trifluoro-1-aminoethyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: 1-(1-Boc-4-piperidyl)-2,2,2-trifluoroethylamine

PC430328-1G
APOSPC430328-1GEA 865 EUR
APOSPC430328-1G
tert-Butyl 4-(2,2,2-trifluoro-1-aminoethyl)piperidine-1-carboxylate
tert-Butyl 4-(2,2,2-trifluoro-1-aminoethyl)piperidine-1-carboxylate
Formel: C₁₂H₂₁F₃N₂O₂
Molecular Weight: 282,303 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12026401
CAS-Nummer: 1159982-64-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR