2,2,2-Trifluoro-1-(4-(4-bromobenzoyl)piperidin-1-yl)ethanone

Lieferant: Apollo Scientific

PC430331-1G
APOSPC430331-1GEA 364 EUR
APOSPC430331-1G
2,2,2-Trifluoro-1-(4-(4-bromobenzoyl)piperidin-1-yl)ethanone
2,2,2-Trifluoro-1-(4-(4-bromobenzoyl)piperidin-1-yl)ethanone
Formel: C₁₄H₁₃BrF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12912608
CAS-Nummer: 203186-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR