2,2,2-Trifluoro-1-(4-benzoylpiperidin-1-yl)ethanone

Lieferant: Apollo Scientific

PC430332-1G
APOSPC430332-1GEA 364 EUR
APOSPC430332-1G
2,2,2-Trifluoro-1-(4-benzoylpiperidin-1-yl)ethanone
2,2,2-Trifluoro-1-(4-benzoylpiperidin-1-yl)ethanone
Formel: C₁₄H₁₄F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00203351
CAS-Nummer: 257946-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR