2,2,2-Trifluoro-1-(4-(4-methylbenzoyl)piperidin-1-yl)ethanone

Lieferant: Apollo Scientific

PC430333-1G
APOSPC430333-1GEA 364 EUR
APOSPC430333-1G
2,2,2-Trifluoro-1-(4-(4-methylbenzoyl)piperidin-1-yl)ethanone
2,2,2-Trifluoro-1-(4-(4-methylbenzoyl)piperidin-1-yl)ethanone
Formel: C₁₅H₁₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12912616
CAS-Nummer: 1198285-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR