1-(4-(2,4-Difluorobenzoyl)piperidin-1-yl)-2,2,2-trifluoroethanone

Lieferant: Apollo Scientific

PC430334-1G
APOSPC430334-1GEA 364 EUR
APOSPC430334-1G
1-(4-(2,4-Difluorobenzoyl)piperidin-1-yl)-2,2,2-trifluoroethanone
1-(4-(2,4-Difluorobenzoyl)piperidin-1-yl)-2,2,2-trifluoroethanone
Formel: C₁₄H₁₂F₅NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12912617
CAS-Nummer: 1198287-09-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR