tert-Butyl 4-(2,4-difluorobenzoyl)piperidine-1-carboxylate

Lieferant: Apollo Scientific

PC430336-1G
APOSPC430336-1GEA 364 EUR
APOSPC430336-1G
tert-Butyl 4-(2,4-difluorobenzoyl)piperidine-1-carboxylate
tert-Butyl 4-(2,4-difluorobenzoyl)piperidine-1-carboxylate
Formel: C₁₇H₂₁F₂NO₃
Molecular Weight: 325,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12068396
CAS-Nummer: 1159825-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR