N-(4-Benzylpiperazin-1-yl)-2,2,2-trifluoroacetamide

Lieferant: Apollo Scientific

PC430339-1G
APOSPC430339-1GEA 546 EUR
APOSPC430339-1G
N-(4-Benzylpiperazin-1-yl)-2,2,2-trifluoroacetamide
N-(4-Benzylpiperazin-1-yl)-2,2,2-trifluoroacetamide
Formel: C₁₃H₁₆F₃N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD12912660
CAS-Nummer: 1198285-47-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR