tert-Butyl 9-hydroxy-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate

Lieferant: Apollo Scientific

PC430343-1G
APOSPC430343-1GEA 865 EUR
APOSPC430343-1G
tert-Butyl 9-hydroxy-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate
tert-Butyl 9-hydroxy-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate
Formel: C₁₄H₂₁F₃N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD28016260
CAS-Nummer: 1823836-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR