tert-Butyl 9-oxo-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate

Lieferant: Apollo Scientific

PC430344-1G
APOSPC430344-1GEA 865 EUR
APOSPC430344-1G
tert-Butyl 9-oxo-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate
tert-Butyl 9-oxo-7-(2,2,2-trifluoroacetyl)-3,7-diaza-bicyclo[3.3.1]nonane-3-carboxylate
Formel: C₁₄H₁₉F₃N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD28016261
CAS-Nummer: 1823420-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR