3-Fluorpyridin-N-oxid
Lieferant: Apollo Scientific
PC430345-5G
APOSPC430345-5GEA
181
EUR
APOSPC430345-5G
3-Fluorpyridin-N-oxid
3-Fluorpyridin-N-oxid
Formel:
C₅H₄FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095181 CAS-Nummer: 695-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...