3-(2'-Fluorbenzyloxy)phenylboronsäure

Lieferant: Apollo Scientific

PC430500-250MG
APOSPC430500-250MGEA 125 EUR
APOSPC430500-250MG
3-(2'-Fluorbenzyloxy)phenylboronsäure
3-(2'-Fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BFO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07369750
CAS-Nummer: 849062-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR