3-Bromo-5-fluoro-2-propoxyphenylboronic acid

Lieferant: Apollo Scientific

Synonyme: (3-Bromo-5-fluoro-2-propoxyphenyl)boronic acid

PC430501-250MG
APOSPC430501-250MGEA 38 EUR
APOSPC430501-250MG
3-Bromo-5-fluoro-2-propoxyphenylboronic acid
3-Bromo-5-fluoro-2-propoxyphenylboronic acid
Formel: C₉H₁₁BBrFO₃
Molecular Weight: 276.895 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07369707
CAS-Nummer: 868272-84-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR