3-(4'-Fluorbenzyloxy)phenylboronsäure

Lieferant: Apollo Scientific

Synonyme: (3-((4-Fluorobenzyl)oxy)phenyl)boronic acid

PC430502-1G
APOSPC430502-1GEA 42.2 EUR
APOSPC430502-1G
3-(4'-Fluorbenzyloxy)phenylboronsäure
3-(4'-Fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246,0420 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08705238
CAS-Nummer: 1072952-03-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR