2,4-Dichloro-5-fluorotoluene
Lieferant: Apollo Scientific
PC430503-2G
APOSPC430503-2GEA
132
EUR
APOSPC430503-2G
2,4-Dichloro-5-fluorotoluene
2,4-Dichloro-5-fluorotoluene
Formel:
C₇H₅Cl₂F Lagertemperatur: Raumtemperatur |
MDL:
MFCD06246852 CAS-Nummer: 86522-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...