Kalium(piperidin-1-ylmethyl)trifluoroborat

Lieferant: Apollo Scientific

PC430504-250MG
APOSPC430504-250MGEA 137 EUR
APOSPC430504-250MG
Kalium(piperidin-1-ylmethyl)trifluoroborat
Kalium(piperidin-1-ylmethyl)trifluoroborat
Formel: C₆H₁₂BF₃N·K
Lagertemperatur: Raumtemperatur
MDL: MFCD10566509
CAS-Nummer: 888711-54-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR