2-Chlor-4-fluor-5-sulfamoylbenzoesäure

Lieferant: Apollo Scientific

PC430506-1G
APOSPC430506-1GEA 119 EUR
APOSPC430506-1G
2-Chlor-4-fluor-5-sulfamoylbenzoesäure
2-Chlor-4-fluor-5-sulfamoylbenzoesäure
Formel: C₇H₅ClFNO₄S
Molecular Weight: 253,6353 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01321357
CAS-Nummer: 4793-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR