5-Fluoro-2-nitrophenylacetonitrile 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(5-Fluoro-2-nitrophenyl)acetonitrile

PC430509-1G
APOSPC430509-1GEA 51 EUR
APOSPC430509-1G
5-Fluoro-2-nitrophenylacetonitrile 95%
5-Fluoro-2-nitrophenylacetonitrile
Formel: C₈H₅FN₂O₂
Molecular Weight: 180,14 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039742
CAS-Nummer: 3456-75-5
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR