4-(4'-Fluorbenzyloxy)phenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: (4-((4-Fluorobenzyl)oxy)phenyl)boronic acid

PC430510-1G
APOSPC430510-1GEA 43.5 EUR
APOSPC430510-1G
4-(4'-Fluorbenzyloxy)phenylboronsäure
4-(4'-Fluorbenzyloxy)phenylboronsäure
Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246.0420 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784403
CAS-Nummer: 871125-82-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR