Potassium 3-trifluoroborato-4-mopholinopropan-1-one

Lieferant: Apollo Scientific

Synonyme: Potassium trifluoro(3-morpholino-3-oxopropyl)borate

PC430512-250MG
APOSPC430512-250MGEA 105 EUR
APOSPC430512-250MG
Potassium 3-trifluoroborato-4-mopholinopropan-1-one
Potassium 3-trifluoroborato-4-mopholinopropan-1-one
Formel: C₇H₁₂BF₃NO₂·K
Molecular Weight: 249.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10700166
CAS-Nummer: 1150654-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR