4-[4-(Trifluoromethyl)phenoxy]piperidine
Lieferant: Apollo Scientific
PC430513-100MG
APOSPC430513-100MGEA
149
EUR
APOSPC430513-100MG
4-[4-(Trifluoromethyl)phenoxy]piperidine
4-[4-(Trifluoromethyl)phenoxy]piperidine
Formel:
C₁₂H₁₄F₃NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD06248882 CAS-Nummer: 28033-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...