4-(Trifluoromethyl)phenethyl alcohol

Lieferant: Apollo Scientific

PC430514-1G
APOSPC430514-1GEA 0 EUR
APOSPC430514-1G
4-(Trifluoromethyl)phenethyl alcohol
4-(Trifluoromethyl)phenethyl alcohol
Formel: C₉H₉F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD06411331
CAS-Nummer: 2968-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR