4-Chloro-7-fluoro-quinoline-3-carbonitrile

Lieferant: Apollo Scientific

PC430515-250MG
APOSPC430515-250MGEA 158 EUR
APOSPC430515-250MG
4-Chloro-7-fluoro-quinoline-3-carbonitrile
4-Chloro-7-fluoro-quinoline-3-carbonitrile
Formel: C₁₀H₄ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06796584
CAS-Nummer: 622369-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR