2-Chlor-6-fluorbenzothiazol

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-6-fluorobenzo[d]thiazole

PC430516-5G
APOSPC430516-5GEA 59.6 EUR
APOSPC430516-5G
2-Chlor-6-fluorbenzothiazol
2-Chloro-6-fluorobenzothiazole
Formel: C₇H₃ClFNS
Molecular Weight: 187,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04448822
CAS-Nummer: 399-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR