2-(2-Fluorophenoxy)-1-ethanamine

Lieferant: Apollo Scientific

PC430517-250MG
APOSPC430517-250MGEA 161 EUR
APOSPC430517-250MG
2-(2-Fluorophenoxy)-1-ethanamine
2-(2-Fluorophenoxy)-1-ethanamine
Formel: C₈H₁₀FNO
Molecular Weight: 155,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00235182
CAS-Nummer: 120351-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR