4-(4-Bromo-2-fluorobenzyl)morpholine
Lieferant: Apollo Scientific
PC430519-1G
APOSPC430519-1GEA
105
EUR
APOSPC430519-1G
4-(4-Bromo-2-fluorobenzyl)morpholine
4-(4-Bromo-2-fluorobenzyl)morpholine
Formel:
C₁₁H₁₃BrFNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD12022322 CAS-Nummer: 338454-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...