2-Chloro-5-trifluoromethyl-benzamidine

Lieferant: Apollo Scientific

PC430521-250MG
APOSPC430521-250MGEA 176 EUR
APOSPC430521-250MG
2-Chloro-5-trifluoromethyl-benzamidine
2-Chloro-5-trifluoromethyl-benzamidine
Formel: C₈H₆ClF₃N₂
Molecular Weight: 222,595 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05662902
CAS-Nummer: 885963-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR