1-(4-Fluorophenyl)homopiperazine
Lieferant: Apollo Scientific
Synonyme:
1-(4-Fluorophenyl)-1,4-diazepane
PC430523-250MG
APOSPC430523-250MGEA
190
EUR
APOSPC430523-250MG
1-(4-Fluorophenyl)homopiperazine
1-(4-Fluorophenyl)homopiperazine
Formel:
C₁₁H₁₅FN₂ Molecular Weight: 194,249 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059822 CAS-Nummer: 751468-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...