4,6-Dichloro-2-(trifluoromethyl)pyrimidin-5-amine

Lieferant: Apollo Scientific

PC430525-100MG
APOSPC430525-100MGEA 200 EUR
APOSPC430525-100MG
4,6-Dichloro-2-(trifluoromethyl)pyrimidin-5-amine
4,6-Dichloro-2-(trifluoromethyl)pyrimidin-5-amine
Formel: C₅H₂Cl₂F₃N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08436613
CAS-Nummer: 2344-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR